Assay Details
Target Gene Details
Entrez Gene ID: | 9414 |
Gene Name: | tight junction protein 2 |
Gene Aliases: |
C9DUPq21.11, DFNA51, DUP9q21.11, PFIC4, X104, ZO2 |
Location: |
Chr.9:69099907-69255208 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TJP2 | XM_017015326.1 | XP_016870815.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508550 | Chr.9:69064250 - 69194517 on Build GRCh38 | Deletion | TJP2 FXN |
nsv6550 | Chr.9:69099228 - 69145132 on Build GRCh38 | Deletion | TJP2 FXN |
esv3891679 | Chr.9:68939243 - 69246082 on Build GRCh38 | Gain | LOC101927069 PRKACG PIP5K1B TJP2 FXN |
esv3576657 | Chr.9:68955164 - 69227475 on Build GRCh38 | Gain | LOC101927069 PRKACG PIP5K1B TJP2 FXN |
dgv7666n100 | Chr.9:68896889 - 69228107 on Build GRCh38 | Gain | LOC101927069 PRKACG PIP5K1B TJP2 FXN |
esv3891678 | Chr.9:68918451 - 69144626 on Build GRCh38 | Gain | LOC101927069 PRKACG PIP5K1B TJP2 FXN |
nsv614548 | Chr.9:68856996 - 69320661 on Build GRCh38 | Gain | BANCR LOC101927069 PRKACG PIP5K1B TJP2 FXN |
dgv7665n100 | Chr.9:68783797 - 69356369 on Build GRCh38 | Gain | BANCR LOC101927069 PRKACG PIP5K1B FAM189A2 TJP2 FXN FAM122A |
nsv466397 | Chr.9:68959841 - 69228107 on Build GRCh38 | Gain | LOC101927069 PRKACG PIP5K1B TJP2 FXN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |