Assay Details
Target Gene Details
Entrez Gene ID: | 11092 |
Gene Name: | sperm acrosome associated 9 |
Gene Aliases: |
C9orf9, Mast |
Location: |
Chr.9:132878320-132890314 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPACA9 | NM_001316897.1 | NP_001303826.1 | ||
NM_001316898.1 | NP_001303827.1 | |||
NM_001316900.1 | NP_001303829.1 | |||
NM_018956.4 | NP_061829.3 | |||
NR_133631.1 | ||||
XM_017014231.1 | XP_016869720.1 | |||
AJ011375.1 | CAB66158.1 | |||
AK075259.1 | BAC11503.1 | |||
BC012940.1 | AAH12940.1 | |||
CK906175.1 | ||||
DA134573.1 | ||||
DA281620.1 | ||||
HY024188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831743 | Chr.9:132820292 - 132982548 on Build GRCh38 | Loss | AK8 MIR548AW GFI1B SPACA9 TSC1 |
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion | MIR3689E LINC00094 STKLD1 COL5A1 SPACA9 WDR5 C9orf62 CELP PPP1R26-AS1 SURF4 MIR3689C OLFM1 FCN2 GTF3C5 MRPS2 ADAMTS13 SURF6 REXO4 GBGT1 SURF2 LOC101448202 MIR3689B MIR6877 DBH-AS1 CEL MIR3689D1 MIR3689F PPP1R26 C9orf116 SNORD141A SLC2A6 LOC101928525 AK8 MED22 LOC105376306 TMEM8C RPL7A DBH CACFD1 FAM163B COL5A1-AS1 SNORD36C SURF1 ABO LOC100130548 BRD3 LOC401557 OBP2B GFI1B MIR4669 SNORD36B MIR3689D2 RNU6ATAC FCN1 SARDH RXRA LOC100996574 ADAMTSL2 VAV2 TSC1 RALGDS SNORD24 SNORD36A MIR548AW LCN1 LOC101928193 MIR3689A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |