Assay Details
Target Gene Details
Entrez Gene ID: | 1621 |
Gene Name: | dopamine beta-hydroxylase |
Gene Aliases: |
DBM |
Location: |
Chr.9:133636363-133659344 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DBH | NM_000787.3 | NP_000778.3 | ||
BC017174.2 | AAH17174.1 | |||
CD014125.1 | ||||
X13255.1 | CAA31631.1 | |||
X13256.1 | CAA31632.1 | |||
Y00096.1 | CAA68285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv615639 | Chr.9:133639476 - 133654580 on Build GRCh38 | Loss |
![]() |
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv951800 | Chr.9:133627979 - 133696678 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
esv3576711 | Chr.9:133582804 - 133778331 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)