Assay Details
Target Gene Details
Entrez Gene ID: | 23586 |
Gene Name: | DEXD/H-box helicase 58 |
Gene Aliases: |
RIG-I, RIGI, RLR-1, SGMRT2 |
Location: |
Chr.9:32455302-32526324 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX58 | NM_014314.3 | NP_055129.2 | ||
AF038963.1 | AAD19826.1 | |||
AK023661.1 | ||||
AK125989.1 | ||||
AK315040.1 | ||||
AL137608.1 | CAB70840.1 | |||
BC132786.1 | ||||
BC136610.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)