Assay Details
Target Gene Details
Entrez Gene ID: | 4712 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B6 |
Gene Aliases: |
B17, CI |
Location: |
Chr.9:32553526-32573184 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFB6 | NM_001199987.1 | NP_001186916.1 | ||
NM_002493.4 | NP_002484.1 | |||
NM_182739.2 | NP_877416.1 | |||
AF035840.1 | AAC68838.1 | |||
AF067167.1 | AAD32451.1 | |||
AK289702.1 | ||||
AW160372.1 | ||||
BC009801.1 | AAH09801.1 | |||
BC029247.1 | AAH29247.1 | |||
BI669356.1 | ||||
BP208846.1 | ||||
BU952486.1 | ||||
CR456920.1 | CAG33201.1 |
Target Gene Details
Entrez Gene ID: | 100129250 |
Gene Name: | TOPORS antisense RNA 1 |
Gene Aliases: |
C9orf133 |
Location: |
Chr.9:32551144-32566860 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOPORS-AS1 | NR_102376.1 | |||
NR_102377.1 | ||||
BI916035.1 | ||||
DA702677.1 | ||||
DA753662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831540 | Chr.9:32509387 - 32651746 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)