Assay Details
Target Gene Details
Entrez Gene ID: | 55664 |
Gene Name: | cell division cycle 37 like 1 |
Gene Aliases: |
CDC37B, HARC |
Location: |
Chr.9:4679553-4708399 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDC37L1 | NM_017913.3 | NP_060383.2 | ||
AK000497.1 | BAA91206.1 | |||
AK000646.1 | BAA91304.1 | |||
AK225510.1 | ||||
BC014133.2 | AAH14133.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1018499 | Chr.9:4665656 - 4706366 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1015376 | Chr.9:4686223 - 4737890 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)