Assay Details
Target Gene Details
Entrez Gene ID: | 8328 |
Gene Name: | growth factor independent 1B transcriptional repressor |
Gene Aliases: |
BDPLT17, ZNF163B |
Location: |
Chr.9:132945545-132991697 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GFI1B | NM_004188.6 | NP_004179.3 | ||
BC043371.1 | AAH43371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509329 | Chr.9:132941822 - 132956074 on Build GRCh38 | Insertion | TSC1 MIR548AW GFI1B |
nsv831743 | Chr.9:132820292 - 132982548 on Build GRCh38 | Loss | SPACA9 TSC1 MIR548AW AK8 GFI1B |
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion | SLC2A6 MIR3689A C9orf116 TSC1 CELP PPP1R26 WDR5 LCN1 MRPS2 SURF2 SNORD141A LOC100996574 LOC105376306 SNORD36B ABO MIR3689B GFI1B SNORD36C MIR3689E SNORD36A LOC100130548 DBH MIR6877 LOC101448202 FCN2 STKLD1 SPACA9 SARDH MIR4669 ADAMTS13 C9orf62 LOC101928525 MIR3689D2 MIR548AW AK8 RNU6ATAC GBGT1 MIR3689C MIR3689F LOC401557 FAM163B DBH-AS1 GTF3C5 VAV2 MED22 RALGDS RPL7A COL5A1-AS1 COL5A1 BRD3 CEL OLFM1 SURF6 SURF4 RXRA MIR3689D1 LINC00094 OBP2B CACFD1 SURF1 SNORD24 PPP1R26-AS1 REXO4 FCN1 LOC101928193 TMEM8C ADAMTSL2 |
nsv6745 | Chr.9:132929207 - 132958453 on Build GRCh38 | Insertion | TSC1 MIR548AW GFI1B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |