Assay Details
Target Gene Details
Entrez Gene ID: | 51104 |
Gene Name: | abhydrolase domain containing 17B |
Gene Aliases: |
C9orf77, CGI-67, FAM108B1 |
Location: |
Chr.9:71859461-71911245 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD17B | NM_001025780.2 | NP_001020951.1 | ||
NM_016014.3 | NP_057098.2 | |||
XM_006717134.3 | XP_006717197.2 | |||
XM_006717135.1 | XP_006717198.1 | |||
XM_017014787.1 | XP_016870276.1 | |||
XM_017014788.1 | XP_016870277.1 | |||
XM_017014789.1 | XP_016870278.1 | |||
XM_017014790.1 | XP_016870279.1 | |||
XM_017014791.1 | XP_016870280.1 | |||
AF151825.1 | AAD34062.1 | |||
AI564777.1 | ||||
AK293060.1 | ||||
BC038390.1 | ||||
BC044576.1 | AAH44576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1050324 | Chr.9:71872257 - 71920574 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)