Assay Details
Target Gene Details
Entrez Gene ID: | 56479 |
Gene Name: | potassium voltage-gated channel subfamily Q member 5 |
Gene Aliases: |
Kv7.5 |
Location: |
Chr.6:72621843-73198851 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNQ5 | NM_001160130.1 | NP_001153602.1 | ||
NM_001160132.1 | NP_001153604.1 | |||
NM_001160133.1 | NP_001153605.1 | |||
NM_001160134.1 | NP_001153606.1 | |||
NM_019842.3 | NP_062816.2 | |||
XM_017011058.1 | XP_016866547.1 | |||
AF202977.1 | AAF69797.1 | |||
AF249278.1 | AAF91335.1 | |||
AF263835.1 | AAF73446.1 | |||
AK299550.1 | ||||
BC050689.1 | AAH50689.1 | |||
BC117359.1 | ||||
BC143554.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)