Assay Details
Target Gene Details
Entrez Gene ID: | 221477 |
Gene Name: | chromosome 6 open reading frame 89 |
Gene Aliases: |
BRAP |
Location: |
Chr.6:36834886-36928964 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C6orf89 | XM_017010434.1 | XP_016865923.1 |
Target Gene Details
Entrez Gene ID: | 51645 |
Gene Name: | peptidylprolyl isomerase like 1 |
Gene Aliases: |
CGI-124, CYPL1, PPIase, hCyPX |
Location: |
Chr.6:36854829-36875024 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPIL1 | NM_016059.4 | NP_057143.1 | ||
AF151882.1 | AAD34119.1 | |||
AK026636.1 | ||||
AY359032.1 | AAQ89391.1 | |||
BC003048.1 | AAH03048.1 | |||
BU195579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5262 | Chr.6:36831553 - 36858434 on Build GRCh38 | Deletion | C6orf89 CPNE5 PPIL1 |
nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain | BTBD9 CDKN1A MTCH1 PPIL1 ZFAND3 DNAH8 PIM1 GLO1 SRSF3 MDGA1 MIR4462 RNF8 LOC107986531 CMTR1 FGD2 RAB44 LOC100131047 TMEM217 GLP1R C6orf89 CCDC167 TBC1D22B CPNE5 LOC100505530 MIR3925 PANDAR PI16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |