Assay Details
Target Gene Details
Entrez Gene ID: | 7915 |
Gene Name: | aldehyde dehydrogenase 5 family member A1 |
Gene Aliases: |
SSADH, SSDH |
Location: |
Chr.6:24494969-24537207 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH5A1 | NM_001080.3 | NP_001071.1 | ||
NM_170740.1 | NP_733936.1 | |||
AJ427353.1 | CAD20882.1 | |||
AJ427354.2 | CAD20883.2 | |||
AJ427355.2 | CAD20884.1 | |||
AK290800.1 | ||||
AK294699.1 | ||||
AK314357.1 | ||||
AK315380.1 | ||||
BC034321.1 | AAH34321.1 | |||
KJ534766.1 | ||||
KJ534767.1 | ||||
KJ534768.1 | ||||
KJ534769.1 | ||||
L34820.1 | AAA67057.1 | |||
Y11192.1 | CAA72076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34020 | Chr.6:24481071 - 24605808 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv519552 | Chr.6:24451028 - 24524476 on Build GRCh38 | Gain |
![]() ![]() |
esv2763542 | Chr.6:24444903 - 24520409 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)