Assay Details
Target Gene Details
Entrez Gene ID: | 285761 |
Gene Name: | discoidin, CUB and LCCL domain containing 1 |
Gene Aliases: |
dJ94G16.1 |
Location: |
Chr.6:117480346-117569858 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCBLD1 | NM_173674.2 | NP_775945.1 | ||
XM_005266941.1 | XP_005266998.1 | |||
XM_006715463.1 | XP_006715526.1 | |||
XM_011535771.1 | XP_011534073.1 | |||
XM_011535773.2 | XP_011534075.1 | |||
XM_011535774.2 | XP_011534076.1 | |||
XM_017010775.1 | XP_016866264.1 | |||
AK095973.1 | BAC04663.1 | |||
BC035671.1 | AAH35671.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)