Assay Details
Target Gene Details
Entrez Gene ID: | 100526836 |
Gene Name: | BLOC1S5-TXNDC5 readthrough (NMD candidate) |
Gene Aliases: |
MUTED-TXNDC5 |
Location: |
Chr.6:7881250-8064414 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BLOC1S5-TXNDC5 | NR_037616.1 | |||
BC052310.1 | AAH52310.1 |
Target Gene Details
Entrez Gene ID: | 81567 |
Gene Name: | thioredoxin domain containing 5 |
Gene Aliases: |
ENDOPDI, ERP46, HCC-2, HCC2, PDIA15, STRF8, UNQ364 |
Location: |
Chr.6:7881250-7910814 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TXNDC5 | NM_001145549.2 | NP_001139021.1 | ||
NM_030810.3 | NP_110437.2 | |||
AJ440721.1 | CAD29430.1 | |||
AK075291.1 | BAC11526.1 | |||
AK098089.1 | ||||
AK315598.1 | ||||
AL833008.1 | CAH56286.1 | |||
AY326464.1 | AAR99514.1 | |||
AY358646.1 | AAQ89009.1 | |||
BC001199.1 | AAH01199.1 | |||
BU855393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv462625 | Chr.6:7868633 - 7906906 on Build GRCh38 | Loss | BMP6 BLOC1S5-TXNDC5 TXNDC5 |
nsv600924 | Chr.6:7823346 - 8206902 on Build GRCh38 | Gain | BMP6 BLOC1S5 PIP5K1P1 EEF1E1 BLOC1S5-TXNDC5 SCARNA27 EEF1E1-BLOC1S5 TXNDC5 |
esv2731544 | Chr.6:7817430 - 8348230 on Build GRCh38 | Deletion | BMP6 BLOC1S5 PIP5K1P1 EEF1E1 BLOC1S5-TXNDC5 SCARNA27 EEF1E1-BLOC1S5 TXNDC5 |
esv3890787 | Chr.6:6996928 - 7981438 on Build GRCh38 | Loss | RIOK1 CAGE1 DSP BMP6 RREB1 SNRNP48 BLOC1S5-TXNDC5 TXNDC5 LOC101928076 SSR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |