Assay Details
Target Gene Details
Entrez Gene ID: | 100507308 |
Gene Name: | TCF21 antisense RNA inducing promoter demethylation |
Gene Aliases: |
EYA4-AS1 |
Location: |
Chr.6:133502252-133889006 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TARID | NR_109982.1 | 6 | 1455 |