Assay Details
Target Gene Details
Entrez Gene ID: | 101927768 |
Gene Name: | uncharacterized LOC101927768 |
Gene Aliases: |
- |
Location: |
Chr.6:113969701-114340729 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927768 | NR_125845.1 |