Assay Details
Target Gene Details
Entrez Gene ID: | 6492 |
Gene Name: | single-minded family bHLH transcription factor 1 |
Gene Aliases: |
bHLHe14 |
Location: |
Chr.6:100385009-100464929 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIM1 | NM_005068.2 | NP_005059.2 | ||
XM_005267100.2 | XP_005267157.1 | |||
XM_011536072.2 | XP_011534374.1 | |||
XM_017011197.1 | XP_016866686.1 | |||
AK308090.1 | ||||
U70212.1 | AAB62395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1028122 | Chr.6:100326369 - 100516471 on Build GRCh38 | Gain | SIM1 ASCC3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |