Assay Details
Target Gene Details
Entrez Gene ID: | 101060264 |
Gene Name: | FOXP4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:41523895-41548621 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP4-AS1 | NR_126415.1 | |||
NR_126416.1 | ||||
NR_126417.1 | 1 | 72 | ||
BF194903.1 | ||||
BG028463.1 | ||||
BQ082169.1 | 1 | 72 | ||
BQ128057.1 | ||||
BQ581966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1073551 | Chr.6:41546161 - 41548062 on Build GRCh38 | Deletion | FOXP4-AS1 FOXP4 |
nsv1144141 | Chr.6:41546162 - 41547362 on Build GRCh38 | Deletion | FOXP4-AS1 FOXP4 |