Assay Details
Target Gene Details
Entrez Gene ID: | 389396 |
Gene Name: | glycine-N-acyltransferase like 3 |
Gene Aliases: |
C6orf140, bA28H17.2 |
Location: |
Chr.6:49499958-49528064 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLYATL3 | NM_001010904.1 | NP_001010904.1 | ||
DB246049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5960n100 | Chr.6:49322472 - 49684411 on Build GRCh38 | Gain | MUT CRISP2 GLYATL3 CENPQ C6orf141 RHAG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |