Assay Details
Target Gene Details
Entrez Gene ID: | 81706 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 14C |
Gene Aliases: |
CPI17-like, KEPI, NY-BR-81 |
Location: |
Chr.6:150143052-150250392 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R14C | NM_030949.2 | NP_112211.1 | ||
AF308297.1 | AAG48264.1 | |||
AF407165.2 | AAL83506.1 | |||
AK096272.1 | ||||
BC015773.1 | AAH15773.1 | |||
CB994599.1 | ||||
DR003369.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3571450 | Chr.6:150163826 - 150180242 on Build GRCh38 | Loss | PPP1R14C |
nsv5536 | Chr.6:150150447 - 150185342 on Build GRCh38 | Insertion | PPP1R14C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |