Assay Details
Target Gene Details
Entrez Gene ID: | 56897 |
Gene Name: | Werner helicase interacting protein 1 |
Gene Aliases: |
WHIP, bA420G6.2 |
Location: |
Chr.6:2765341-2785745 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRNIP1 | NM_020135.2 | NP_064520.2 | ||
NM_130395.2 | NP_569079.1 | |||
AB056152.1 | BAB60709.1 | |||
AB209723.1 | BAD92960.1 | |||
AF218313.1 | AAF80563.1 | |||
AK026179.1 | BAB15383.1 | |||
AK094823.1 | ||||
AK223593.1 | BAD97313.1 | |||
AK315047.1 | ||||
AK315471.1 | ||||
BC018923.2 | AAH18923.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)