Assay Details
Target Gene Details
Entrez Gene ID: | 100508120 |
Gene Name: | GMDS antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.6:2245753-2413591 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GMDS-AS1 | NR_046229.1 | |||
CN395927.1 | ||||
CR739457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3607900 | Chr.6:2093592 - 2775672 on Build GRCh38 | Gain | LINC01600 WRNIP1 GMDS MYLK4 GMDS-AS1 |
esv2763526 | Chr.6:2229830 - 2311682 on Build GRCh38 | Loss | GMDS GMDS-AS1 |