Assay Details
Target Gene Details
Entrez Gene ID: | 639 |
Gene Name: | PR domain 1 |
Gene Aliases: |
BLIMP1, PRDI-BF1 |
Location: |
Chr.6:106046729-106109939 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRDM1 | XM_017011187.1 | XP_016866676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1025231 | Chr.6:106003137 - 106186514 on Build GRCh38 | Gain | PRDM1 ATG5 |
esv3610264 | Chr.6:106047896 - 106058621 on Build GRCh38 | Loss | PRDM1 |
esv3610263 | Chr.6:106046710 - 106062130 on Build GRCh38 | Loss | PRDM1 |
nsv830755 | Chr.6:106013830 - 106176327 on Build GRCh38 | Loss | PRDM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |