Assay Details
Target Gene Details
Entrez Gene ID: | 112744 |
Gene Name: | interleukin 17F |
Gene Aliases: |
CANDF6, IL-17F, ML-1, ML1 |
Location: |
Chr.6:52236681-52245689 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL17F | NM_052872.3 | NP_443104.1 | ||
XM_011514276.1 | XP_011512578.1 | |||
AF384857.1 | AAK83350.1 | |||
BC070124.1 | AAH70124.1 | |||
JF305974.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830662 | Chr.6:52064873 - 52264478 on Build GRCh38 | Gain | MIR133B MIR206 IL17F IL17A MCM3 PKHD1 LINCMD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |