Assay Details
Target Gene Details
Entrez Gene ID: | 221391 |
Gene Name: | opsin 5 |
Gene Aliases: |
GPR136, GRP136, PGR12, TMEM13 |
Location: |
Chr.6:47782037-47831088 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OPN5 | NM_181744.3 | NP_859528.1 | ||
XM_017010416.1 | XP_016865905.1 | |||
AY288419.1 | AAP72128.1 | |||
AY377391.1 | AAR21109.1 | |||
BC126194.1 | ||||
BC126198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain | TNFRSF21 ADGRF1 LOC101926962 PTCHD4 ADGRF2 CD2AP ADGRF4 ADGRF5 OPN5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |