Assay Details
Target Gene Details
Entrez Gene ID: | 100507462 |
Gene Name: | GVQW motif containing 2 |
Gene Aliases: |
- |
Location: |
Chr.6:138725211-138773679 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GVQW2 | NM_001242740.1 | NP_001229669.1 | ||
AK092592.1 | ||||
DN917422.1 |