Assay Details
Target Gene Details
Entrez Gene ID: | 55862 |
Gene Name: | ethylmalonyl-CoA decarboxylase 1 |
Gene Aliases: |
HEL-S-76, MMCD, dJ351K20.2 |
Location: |
Chr.6:127288712-127343609 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ECHDC1 | NM_001002030.1 | NP_001002030.1 | ||
NM_001105544.1 | NP_001099014.1 | |||
NM_001105545.1 | NP_001099015.1 | |||
NM_001139510.1 | NP_001132982.1 | |||
NM_018479.3 | NP_060949.2 | |||
XM_005267047.2 | XP_005267104.1 | |||
XM_005267048.1 | XP_005267105.1 | |||
XM_005267049.2 | XP_005267106.1 | |||
XM_005267050.2 | XP_005267107.1 | |||
XM_011535942.2 | XP_011534244.1 | |||
XM_017011056.1 | XP_016866545.1 | |||
AF220192.1 | AAF67657.1 | |||
AK021464.1 | ||||
AK025760.1 | ||||
AK025796.1 | ||||
AK098146.1 | ||||
AK303812.1 | ||||
AL834469.1 | CAD39128.1 | |||
BC003549.1 | AAH03549.1 | |||
BE896631.1 | ||||
BQ224769.1 | ||||
DA846978.1 | ||||
EU668355.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |