Assay Details
Target Gene Details
Entrez Gene ID: | 2070 |
Gene Name: | EYA transcriptional coactivator and phosphatase 4 |
Gene Aliases: |
CMD1J, DFNA10 |
Location: |
Chr.6:133240340-133532120 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EYA4 | NM_001301012.1 | NP_001287941.1 | ||
NM_001301013.1 | NP_001287942.1 | |||
NM_004100.4 | NP_004091.3 | |||
NM_172103.3 | NP_742101.2 | |||
NM_172105.3 | NP_742103.1 | |||
XM_005266851.4 | XP_005266908.1 | |||
XM_005266853.4 | XP_005266910.1 | |||
XM_017010368.1 | XP_016865857.1 | |||
XM_017010369.1 | XP_016865858.1 | |||
XM_017010370.1 | XP_016865859.1 | |||
XM_017010371.1 | XP_016865860.1 | |||
XM_017010372.1 | XP_016865861.1 | |||
XM_017010373.1 | XP_016865862.1 | |||
XM_017010374.1 | XP_016865863.1 | |||
XM_017010375.1 | XP_016865864.1 | |||
AK295798.1 | ||||
AK299378.1 | ||||
AK301950.1 | ||||
BC014193.2 | AAH14193.2 | |||
BC041063.1 | AAH41063.1 | |||
KU178044.1 | ||||
KU178045.1 | ||||
Y17114.1 | CAA76636.1 |
Target Gene Details
Entrez Gene ID: | 100507308 |
Gene Name: | TCF21 antisense RNA inducing promoter demethylation |
Gene Aliases: |
EYA4-AS1 |
Location: |
Chr.6:133502252-133889006 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TARID | NR_109982.1 | |||
AK093513.1 | ||||
BQ009762.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |