Assay Details
Target Gene Details
Entrez Gene ID: | 221476 |
Gene Name: | peptidase inhibitor 16 |
Gene Aliases: |
CD364, CRISP9, MSMBBP, PSPBP |
Location: |
Chr.6:36948263-36964837 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PI16 | NM_001199159.1 | NP_001186088.1 | ||
NM_153370.2 | NP_699201.2 | |||
XM_005248917.2 | XP_005248974.1 | |||
XM_011514375.2 | XP_011512677.1 | |||
XM_017010430.1 | XP_016865919.1 | |||
AK075470.1 | BAC11640.1 | |||
AK124589.1 | BAC85892.1 | |||
AK302193.1 | ||||
AK303928.1 | ||||
AK312785.1 | ||||
AY358422.1 | AAQ88788.1 | |||
BC022399.1 | AAH22399.2 | |||
BC035634.1 | ||||
DA342875.1 | ||||
DB178215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv380n21 | Chr.6:36948130 - 36957998 on Build GRCh38 | Gain | PI16 |
nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain | TBC1D22B CDKN1A CMTR1 MTCH1 DNAH8 LOC100505530 C6orf89 ZFAND3 FGD2 PANDAR MDGA1 TMEM217 RAB44 CPNE5 LOC100131047 CCDC167 GLP1R BTBD9 RNF8 PPIL1 MIR4462 LOC107986531 GLO1 PIM1 SRSF3 PI16 MIR3925 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |