Assay Details
Target Gene Details
Entrez Gene ID: | 221477 |
Gene Name: | chromosome 6 open reading frame 89 |
Gene Aliases: |
BRAP |
Location: |
Chr.6:36834886-36928964 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C6orf89 | XM_017010434.1 | XP_016865923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5262 | Chr.6:36831553 - 36858434 on Build GRCh38 | Deletion | PPIL1 CPNE5 C6orf89 |
nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain | CCDC167 RAB44 MIR3925 DNAH8 GLP1R CMTR1 MIR4462 PPIL1 ZFAND3 GLO1 RNF8 C6orf89 MTCH1 MDGA1 CPNE5 PI16 PIM1 BTBD9 LOC100131047 SRSF3 CDKN1A LOC100505530 TMEM217 PANDAR FGD2 LOC107986531 TBC1D22B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |