Assay Details
Target Gene Details
Entrez Gene ID: | 51696 |
Gene Name: | hdc homolog, cell cycle regulator |
Gene Aliases: |
HDC, HDCL, HHDC, dJ225E12.1 |
Location: |
Chr.6:139135112-139180809 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HECA | NM_016217.2 | NP_057301.1 | ||
AB033492.1 | BAA85322.1 | |||
BC070068.1 | AAH70068.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |