Assay Details
Target Gene Details
Entrez Gene ID: | 101928191 |
Gene Name: | uncharacterized LOC101928191 |
Gene Aliases: |
- |
Location: |
Chr.6:10932317-10954089 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928191 | NR_125851.1 |
Target Gene Details
Entrez Gene ID: | 221711 |
Gene Name: | synaptonemal complex protein 2 like |
Gene Aliases: |
C6orf177, NO145, dJ62D2.1 |
Location: |
Chr.6:10886831-10974309 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SYCP2L | NM_001040274.2 | NP_001035364.2 | ||
AK128130.1 | BAC87288.1 | |||
AM293674.1 | ||||
BC012225.1 | AAH12225.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |