Assay Details
Target Gene Details
Entrez Gene ID: | 9457 |
Gene Name: | four and a half LIM domains 5 |
Gene Aliases: |
1700027G07Rik, ACT, FHL-5, dJ393D12.2 |
Location: |
Chr.6:96562525-96618626 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FHL5 | NM_001170807.2 | NP_001164278.1 | ||
NM_001322466.1 | NP_001309395.1 | |||
NM_001322467.1 | NP_001309396.1 | |||
NM_020482.5 | NP_065228.4 | |||
AF278541.1 | AAF85978.1 | |||
AK057611.1 | ||||
AK058175.1 | ||||
AK223539.1 | BAD97259.1 | |||
AK314608.1 | ||||
BC021723.2 | AAH21723.1 | |||
BC029490.1 | AAH29490.1 | |||
DA099831.1 | ||||
DA234984.1 | ||||
HY020942.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)