Assay Details
Target Gene Details
Entrez Gene ID: | 3360 |
Gene Name: | 5-hydroxytryptamine receptor 4 |
Gene Aliases: |
5-HT4, 5-HT4R |
Location: |
Chr.5:148451032-148654527 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HTR4 | NM_001040169.2 | NP_001035259.1 | ||
NM_199453.3 | NP_955525.1 | |||
AJ011371.1 | CAA09600.1 | |||
AJ131725.1 | CAC79533.1 | |||
AJ278979.1 | CAC22248.1 | |||
AJ278981.1 | CAC22250.1 | |||
AJ633645.1 | CAG17627.1 | |||
Y08756.1 | CAA70002.1 | |||
Y09586.1 | CAA70774.1 | |||
Y13584.1 | CAA73911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830516 | Chr.5:148421541 - 148586530 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)