Assay Details
Target Gene Details
Entrez Gene ID: | 2149 |
Gene Name: | coagulation factor II thrombin receptor |
Gene Aliases: |
CF2R, HTR, PAR-1, PAR1, TR |
Location: |
Chr.5:76716043-76735780 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
F2R | NM_001311313.1 | NP_001298242.1 | ||
NM_001992.4 | NP_001983.2 | |||
AK297131.1 | ||||
AK308169.1 | ||||
AK314202.1 | ||||
BC002464.2 | AAH02464.1 | |||
BC051909.1 | AAH51909.1 | |||
BT007279.1 | AAP35943.1 | |||
DA799420.1 | ||||
M62424.1 | AAA36743.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035135 | Chr.5:76631202 - 76796233 on Build GRCh38 | Gain | F2R IQGAP2 NCRUPAR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |