Assay Details
Target Gene Details
Entrez Gene ID: | 11044 |
Gene Name: | PAP associated domain containing 7 |
Gene Aliases: |
LAK-1, LAK1, POLK, POLS, TRF4, TRF4-1, TRF41, TUTASE5 |
Location: |
Chr.5:6713952-6757048 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAPD7 | NM_001171805.1 | NP_001165276.1 | ||
NM_006999.4 | NP_008930.1 | |||
XM_005248234.3 | XP_005248291.1 | |||
XM_017008989.1 | XP_016864478.1 | |||
AF089896.1 | AAD45198.1 | |||
AI871781.1 | ||||
BC084567.1 | ||||
KC424495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv596947 | Chr.5:6680002 - 6744685 on Build GRCh38 | Loss |
![]() ![]() |
esv3603920 | Chr.5:6719857 - 6733105 on Build GRCh38 | Loss |
![]() |
nsv822994 | Chr.5:6701714 - 6766012 on Build GRCh38 | Gain |
![]() ![]() |
nsv596949 | Chr.5:6705744 - 6777656 on Build GRCh38 | Loss |
![]() ![]() |
nsv822993 | Chr.5:6689024 - 6753962 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)