Assay Details
Target Gene Details
Entrez Gene ID: | 100861519 |
Gene Name: | GDNF antisense RNA 1 (head to head) |
Gene Aliases: |
GDNFOS |
Location: |
Chr.5:37840028-37875799 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GDNF-AS1 | NR_103441.1 | |||
XM_017008916.1 | XP_016864405.1 | |||
XM_017008918.1 | XP_016864407.1 | |||
JF824130.1 |