Assay Details
Target Gene Details
Entrez Gene ID: | 1182 |
Gene Name: | chloride voltage-gated channel 3 |
Gene Aliases: |
CLC3, ClC-3 |
Location: |
Chr.4:169620521-169723187 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLCN3 | NM_001243372.1 | NP_001230301.1 | ||
NM_001829.3 | NP_001820.2 | |||
NM_173872.3 | NP_776297.2 | |||
AB019542.1 | BAC54560.1 | |||
AF029346.1 | AAB95161.1 | |||
AF172729.1 | AAD51034.1 | |||
AK304362.1 | ||||
BC054877.1 | AAH54877.1 | |||
BC136510.1 | ||||
BX647119.1 | ||||
X78520.1 | CAA55280.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |