Assay Details
Target Gene Details
Entrez Gene ID: | 201973 |
Gene Name: | primase and DNA directed polymerase |
Gene Aliases: |
CCDC111, MYP22 |
Location: |
Chr.4:184649613-184694959 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2763423 | Chr.4:184642465 - 184688651 on Build GRCh38 | Loss | PRIMPOL CASP3 |
nsv830168 | Chr.4:184626297 - 184801578 on Build GRCh38 | Loss | PRIMPOL CENPU SLED1 ACSL1 CASP3 |
nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain | LVCAT8 PRIMPOL CENPU ACSL1 IRF2 CASP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |