Assay Details
Target Gene Details
Entrez Gene ID: | 55768 |
Gene Name: | N-glycanase 1 |
Gene Aliases: |
CDDG, CDG1V, PNG1, PNGase |
Location: |
Chr.3:25718944-25790039 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NGLY1 | NM_001145293.1 | NP_001138765.1 | ||
NM_001145294.1 | NP_001138766.1 | |||
NM_001145295.1 | NP_001138767.1 | |||
NM_018297.3 | NP_060767.2 | |||
XM_005265316.1 | XP_005265373.1 | |||
XM_005265317.1 | XP_005265374.1 | |||
XM_011533944.1 | XP_011532246.1 | |||
XM_017006839.1 | XP_016862328.1 | |||
AB209549.1 | BAD92786.1 | |||
AF250924.2 | AAF74720.2 | |||
AK001867.1 | BAA91950.1 | |||
AK296047.1 | ||||
AK310473.1 | ||||
AL832748.1 | ||||
BC000963.2 | AAH00963.1 | |||
BC007226.1 | AAH07226.1 | |||
BC017220.2 | AAH17220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893676 | Chr.3:24984204 - 26577498 on Build GRCh38 | Loss | TOP2B NGLY1 LOC105376997 LINC00692 MIR4442 RARB OXSM |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |