Assay Details
Target Gene Details
Entrez Gene ID: | 10242 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 2 |
Gene Aliases: |
- |
Location: |
Chr.3:178536298-178844429 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNMB2 | NM_181361.2 | NP_852006.1 | ||
XM_011512325.2 | XP_011510627.1 | |||
AK298291.1 | ||||
BC017825.1 | AAH17825.1 | |||
BG701449.1 | ||||
DA532966.1 |
Target Gene Details
Entrez Gene ID: | 104797538 |
Gene Name: | KCNMB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:178525467-178860405 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNMB2-AS1 | NR_126560.1 | |||
NR_126561.1 | ||||
AF279780.1 | ||||
AY769439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524447 | Chr.3:178539801 - 178638481 on Build GRCh38 | Gain | KCNMB2-AS1 KCNMB2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |