Assay Details
Target Gene Details
Entrez Gene ID: | 6529 |
Gene Name: | solute carrier family 6 member 1 |
Gene Aliases: |
GABATHG, GABATR, GAT1, MAE |
Location: |
Chr.3:10992718-11039249 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC6A1 | NM_003042.3 | NP_003033.3 | ||
XM_006713306.3 | XP_006713369.1 | |||
BC033904.1 | AAH33904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | LINC00852 LINC00606 ATG7 SLC6A1 GHRLOS TAMM41 SLC6A1-AS1 TATDN2 ATP2B2 IRAK2 FANCD2OS HRH1 SLC6A11 BRK1 VHL FANCD2 SEC13 LOC101927467 ATP2B2-IT2 MIR885 MIR378B GHRL VGLL4 |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | LOC401052 LINC00606 ATG7 SLC6A1 SLC6A1-AS1 TATDN2 ATP2B2 IRAK2 VHL FANCD2 EMC3 ATP2B2-IT2 MIR885 CIDECP LINC00852 GHRLOS TAMM41 FANCD2OS HRH1 SLC6A11 BRK1 SEC13 FANCD2P2 LOC101927467 MIR378B GHRL VGLL4 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | LINC00852 LINC00606 ATG7 SLC6A1 GHRLOS TAMM41 SLC6A1-AS1 TATDN2 ATP2B2 IRAK2 FANCD2OS HRH1 SLC6A11 BRK1 VHL FANCD2 SEC13 FANCD2P2 LOC101927467 ATP2B2-IT2 MIR885 MIR378B GHRL VGLL4 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | LINC00852 LINC00606 ATG7 SLC6A1 GHRLOS TAMM41 SLC6A1-AS1 TATDN2 ATP2B2 IRAK2 FANCD2OS HRH1 SLC6A11 BRK1 VHL FANCD2 SEC13 FANCD2P2 LOC101927467 ATP2B2-IT2 MIR885 MIR378B GHRL VGLL4 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | LOC401052 LINC00606 ATG7 SLC6A1 SLC6A1-AS1 TATDN2 ATP2B2 IRAK2 VHL FANCD2 EMC3-AS1 EMC3 ATP2B2-IT2 MIR885 CIDECP LINC00852 GHRLOS TAMM41 FANCD2OS HRH1 SLC6A11 BRK1 SEC13 FANCD2P2 LOC101927467 MIR378B GHRL VGLL4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |