Assay Details
Target Gene Details
Entrez Gene ID: | 100862662 |
Gene Name: | ALDH1L1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.3:126180065-126210169 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH1L1-AS2 | NR_046383.1 | |||
DA256650.1 | ||||
DA566603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3985 | Chr.3:126173880 - 126218546 on Build GRCh38 | Deletion | ALDH1L1 ALDH1L1-AS2 |
dgv8545n54 | Chr.3:126091128 - 126251891 on Build GRCh38 | Loss | ALDH1L1-AS1 SLC41A3 ALDH1L1 ALDH1L1-AS2 |
nsv1007256 | Chr.3:126163635 - 126238737 on Build GRCh38 | Gain | ALDH1L1 ALDH1L1-AS2 |