Assay Details
Target Gene Details
Entrez Gene ID: | 1371 |
Gene Name: | coproporphyrinogen oxidase |
Gene Aliases: |
CPO, CPX, HCP |
Location: |
Chr.3:98568448-98593684 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPOX | NM_000097.5 | NP_000088.3 | ||
XM_005247125.4 | XP_005247182.1 | |||
AK223481.1 | BAD97201.1 | |||
AK290140.1 | ||||
BC017210.2 | AAH17210.1 | |||
BC023551.2 | AAH23551.1 | |||
BC023554.2 | AAH23554.1 | |||
D16611.1 | BAA04033.1 | |||
Z28409.1 | CAA82250.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834775 | Chr.3:98451215 - 98648708 on Build GRCh38 | Gain | OR5K2 CLDND1 GPR15 OR5K1 CPOX |
nsv591065 | Chr.3:98485354 - 99047482 on Build GRCh38 | Gain | OR5K2 ST3GAL6-AS1 DCBLD2 CLDND1 GPR15 ST3GAL6 CPOX |
dgv293n21 | Chr.3:98382257 - 98598992 on Build GRCh38 | Loss | OR5K2 CLDND1 OR5K3 GPR15 OR5K1 CPOX |
dgv4819n100 | Chr.3:98565439 - 98662629 on Build GRCh38 | Gain | CPOX |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |