Assay Details
Target Gene Details
Entrez Gene ID: | 6259 |
Gene Name: | receptor-like tyrosine kinase |
Gene Aliases: |
D3S3195, JTK5, JTK5A, RYK1 |
Location: |
Chr.3:134157133-134250834 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RYK | NM_001005861.2 | NP_001005861.1 | ||
NM_002958.3 | NP_002949.2 | |||
XM_017007016.1 | XP_016862505.1 | |||
AB209405.1 | BAD92642.1 | |||
BC021700.2 | AAH21700.2 | |||
S59184.1 | AAB26341.1 | |||
X69970.1 | CAA49591.1 | |||
X96588.1 | CAA65406.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)