Assay Details
Target Gene Details
Entrez Gene ID: | 100505385 |
Gene Name: | IQCJ-SCHIP1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.3:159069252-159897366 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IQCJ-SCHIP1 | NM_001197113.1 | NP_001184042.1 | ||
NM_001197114.1 | NP_001184043.1 | |||
AK096479.1 | ||||
DQ157847.1 | ||||
DQ157848.1 |
Target Gene Details
Entrez Gene ID: | 29970 |
Gene Name: | schwannomin interacting protein 1 |
Gene Aliases: |
SCHIP-1 |
Location: |
Chr.3:159273247-159897366 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCHIP1 | NM_001197107.1 | NP_001184036.1 | ||
NM_001197108.1 | NP_001184037.1 | |||
NM_014575.3 | NP_055390.1 | |||
AF145713.1 | AAF34241.1 | |||
AF145714.1 | AAF34242.1 | |||
AF145715.1 | AAF34243.1 | |||
AK314662.1 | ||||
BC036535.2 | AAH36535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470971 | Chr.3:158873139 - 159440428 on Build GRCh38 | Gain | LOC105374181 MIR3919 SCHIP1 IQCJ-SCHIP1 IQCJ |
nsv998949 | Chr.3:159057017 - 159676761 on Build GRCh38 | Gain | MIR3919 SCHIP1 IQCJ-SCHIP1 IQCJ |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |