Assay Details
Target Gene Details
Entrez Gene ID: | 788 |
Gene Name: | solute carrier family 25 member 20 |
Gene Aliases: |
CAC, CACT |
Location: |
Chr.3:48856923-48898993 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A20 | NM_000387.5 | NP_000378.1 | ||
AK301744.1 | ||||
AK312962.1 | ||||
BC001689.2 | AAH01689.1 | |||
BI913588.1 | ||||
DC360427.1 | ||||
Y10319.1 | CAA71367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893694 | Chr.3:48792101 - 48921349 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)