Assay Details
Target Gene Details
Entrez Gene ID: | 51099 |
Gene Name: | abhydrolase domain containing 5 |
Gene Aliases: |
CDS, CGI58, IECN2, NCIE2 |
Location: |
Chr.3:43690883-43722725 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD5 | NM_016006.4 | NP_057090.2 | ||
XM_011533779.2 | XP_011532081.1 | |||
XM_011533780.1 | XP_011532082.1 | |||
AF151816.1 | AAD34053.1 | |||
AK313811.1 | ||||
BC021958.1 | AAH21958.1 | |||
BG721387.1 | ||||
KU178625.1 | ||||
KU178626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834673 | Chr.3:43581493 - 43758384 on Build GRCh38 | Gain | ABHD5 ANO10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |