Assay Details
Target Gene Details
Entrez Gene ID: | 152404 |
Gene Name: | immunoglobulin superfamily member 11 |
Gene Aliases: |
BT-IgSF, CT119, CXADRL1, Igsf13, VSIG3 |
Location: |
Chr.3:118900630-119146068 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3960 | Chr.3:118883470 - 118914525 on Build GRCh38 | Insertion | IGSF11 |
nsv1011135 | Chr.3:116380194 - 119036472 on Build GRCh38 | Gain | LINC00901 LSAMP LOC105374060 IGSF11 MIR4447 TUSC7 IGSF11-AS1 LOC101926968 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |