Assay Details
Target Gene Details
Entrez Gene ID: | 201626 |
Gene Name: | phosphodiesterase 12 |
Gene Aliases: |
2'-PDE, 2-PDE |
Location: |
Chr.3:57556247-57656480 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDE12 | NM_001322176.1 | NP_001309105.1 | ||
BM713922.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv590368 | Chr.3:57483370 - 57576753 on Build GRCh38 | Gain | ARF4 DNAH12 PDE12 |
nsv1008684 | Chr.3:57537986 - 57766490 on Build GRCh38 | Gain | DENND6A ARF4 DNAH12 LOC101929159 SLMAP PDE12 ARF4-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |