Assay Details
Target Gene Details
Entrez Gene ID: | 51726 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B11 |
Gene Aliases: |
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537 |
Location: |
Chr.3:186570676-186585800 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNAJB11 | NM_016306.5 | NP_057390.1 | ||
AB028859.2 | BAA88307.1 | |||
AF228505.1 | AAF61711.1 | |||
AF277317.1 | AAK69110.1 | |||
AJ250137.1 | CAB65118.1 | |||
AK075300.1 | BAC11533.1 | |||
AK075430.1 | BAC11617.1 | |||
AK301292.1 | ||||
AY359043.1 | AAQ89402.1 | |||
BC001144.1 | AAH01144.1 | |||
BM754506.1 | ||||
BT007063.1 | AAP35712.1 | |||
CR457096.1 | CAG33377.1 | |||
DA058195.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |